4-(4-Nitrophenoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

169977-1G 169977-500MG 169977-5G 169977-25G
77465-780EA 415.09 USD
77465-780 77478-534 77488-732 77499-250
4-(4-Nitrophenoxy)-1,1'-biphenyl ≥97%
4-(4-Nitrophenoxy)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₈H₁₃NO₃
MW: 291.3007 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00222026
CAS Number: 6641-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR