4-Iodo-2-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

169943-1G 169943-500MG 169943-5G 169943-25G
77465-776EA 409.94 USD
77465-776 77478-530 77488-728 77499-246
4-Iodo-2-isopropylphenol ≥97%
4-Iodo-2-isopropylphenol

Precaución: For research use only

Formula: C₉H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD16997105
CAS Number: 1243439-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR