(3-(phenoxycarbonyl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

169775-1G 169775-500MG 169775-5G 169775-25G
77465-772EA 415.09 USD
77465-772 77478-526 77488-724 77499-242
(3-(phenoxycarbonyl)phenyl)boronic acid ≥97%
(3-(phenoxycarbonyl)phenyl)boronic acid

Precaución: For research use only

Formula: C₁₃H₁₁BO₄
MW: 242.0350 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852934
CAS Number: 2051379-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR