5-Chloro-2-(ethylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-(ethylthio)pyridine

169389-1G 169389-500MG 169389-5G 169389-25G
77465-764EA 328.57 USD
77465-764 77478-520 77488-712 77499-230
5-Chloro-2-(ethylthio)pyridine ≥97%
5-Chloro-2-(ethylthio)pyridine

Precaución: For research use only

Formula: C₇H₈ClNS
MW: 173.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21641935
CAS Number: 102645-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR