3-Benzyloxy-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16914-1G 16914-500MG 16914-5G 16914-25G
77465-760EA 109.18 USD
77465-760 77478-516 77488-708 77499-226
3-Benzyloxy-4-fluorobenzaldehyde ≥97%
3-Benzyloxy-4-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₄H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11110161
CAS Number: 103438-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR