5-Bromo-4-methyl-3-thiophenecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

168813-1G 168813-500MG 168813-5G
77465-756EA 633.45 USD
77465-756 77478-512 77488-704
5-Bromo-4-methyl-3-thiophenecarboxylic acid ≥97%
5-Bromo-4-methyl-3-thiophenecarboxylic acid

Precaución: For research use only

Formula: C₆H₅BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD24107252
CAS Number: 1523182-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR