4-(Ethylthio)-α,α-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168766-1G 168766-500MG 168766-5G
77465-752EA 648.9 USD
77465-752 77478-508 77488-700
4-(Ethylthio)-α,α-dimethylbenzenemethanol ≥95%
4-(Ethylthio)-α,α-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD20528505
CAS Number: 1379317-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR