α,α-Dimethyl-4-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168736-1G 168736-500MG 168736-5G
77465-748EA 648.9 USD
77465-748 77478-504 77488-696
α,α-Dimethyl-4-(methylthio)benzenemethanol ≥95%
α,α-Dimethyl-4-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₄OS
Storage Temperature: Refrigerator
MDL Number: MFCD20528506
CAS Number: 82955-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR