4-(1,1-Dimethylethyl)-α,α-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168717-1G 168717-500MG 168717-5G
77465-744EA 668.47 USD
77465-744 77478-500 77488-692
4-(1,1-Dimethylethyl)-α,α-dimethylbenzenemethanol ≥95%
4-(1,1-Dimethylethyl)-α,α-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD00029998
CAS Number: 23853-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR