α,α-Dimethyl-2-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168693-1G 168693-500MG 168693-5G 168693-25G
77465-740EA 246.17 USD
77465-740 77478-496 77488-688 77499-218
α,α-Dimethyl-2-(1-methylethyl)benzenemethanol ≥95%
α,α-Dimethyl-2-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD09965967
CAS Number: 106989-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR