α,α-Dimethyl-3-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168674-1G 168674-500MG 168674-5G 168674-25G
77465-736EA 210.12 USD
77465-736 77478-492 77488-682 77499-214
α,α-Dimethyl-3-(1-methylethyl)benzenemethanol ≥95%
α,α-Dimethyl-3-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Refrigerator
MDL Number: MFCD00046571
CAS Number: 14860-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR