4-(1-Hydroxyethyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

168655-1G 168655-500MG 168655-5G 168655-25G
77465-732EA 109.18 USD
77465-732 77478-488 77488-678 77499-212
4-(1-Hydroxyethyl)benzonitrile ≥97%
4-(1-Hydroxyethyl)benzonitrile

Precaución: For research use only

Formula: C₉H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00870635
CAS Number: 52067-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR