4-(Ethylthio)-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168634-1G 168634-500MG 168634-5G 168634-25G
77465-728EA 341.96 USD
77465-728 77478-484 77488-674 77499-208
4-(Ethylthio)-α-methylbenzenemethanol ≥95%
4-(Ethylthio)-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16300492
CAS Number: 1225470-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR