2-(2,6-Dimethylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

168569-1G 168569-500MG 168569-5G 168569-25G
77465-724EA 390.37 USD
77465-724 77478-480 77488-670 77499-206
2-(2,6-Dimethylphenyl)cyclohexanol ≥95%
2-(2,6-Dimethylphenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₄H₂₀O
Storage Temperature: Ambient
CAS Number: 2737207-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR