1-(Ethylthio)-4-pentylbenzene ≥95%

Supplier: AOBChem USA

Warning

168031-1G 168031-500MG 168031-5G 168031-25G
77465-712EA 415.09 USD
77465-712 77478-468 77488-656 77499-196
1-(Ethylthio)-4-pentylbenzene ≥95%
1-(Ethylthio)-4-pentylbenzene

Precaución: For research use only

Formula: C₁₃H₂₀S
MW: 208.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404451
CAS Number: 2737206-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR