1-(Ethylthio)-3-(mEthylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

168027-1G 168027-500MG 168027-5G
77465-708EA 648.9 USD
77465-708 77478-464 77488-652
1-(Ethylthio)-3-(mEthylthio)benzene ≥95%
1-(Ethylthio)-3-(mEthylthio)benzene

Precaución: For research use only

Formula: C₉H₁₂S₂
MW: 184.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617729
CAS Number: 83500-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR