1-Ethoxy-3-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

168025-1G 168025-500MG 168025-5G 168025-25G
77465-704EA 367.71 USD
77465-704 77478-460 77488-648 77499-190
1-Ethoxy-3-(ethylthio)benzene ≥95%
1-Ethoxy-3-(ethylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD11617731
CAS Number: 85428-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR