1-Ethyl-3-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

168016-1G 168016-500MG 168016-5G
77465-700EA 668.47 USD
77465-700 77478-456 77488-644
1-Ethyl-3-(ethylthio)benzene ≥95%
1-Ethyl-3-(ethylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617725
CAS Number: 158321-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR