2-(Ethylthio)-N,N-dimethylbenzenamine ≥95%

Supplier: AOBChem USA

Warning

167990-1G 167990-500MG 167990-5G 167990-25G
77465-696EA 415.09 USD
77465-696 77478-452 77488-640 77499-188
2-(Ethylthio)-N,N-dimethylbenzenamine ≥95%
2-(Ethylthio)-N,N-dimethylbenzenamine

Precaución: For research use only

Formula: C₁₀H₁₅NS
MW: 181.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852408
CAS Number: 87094-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR