1-(Ethylthio)-2,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

167986-1G 167986-500MG 167986-5G
77465-692EA 648.9 USD
77465-692 77478-448 77488-636
1-(Ethylthio)-2,3-dimethylbenzene ≥95%
1-(Ethylthio)-2,3-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617737
CAS Number: 105371-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR