1-Ethyl-4-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

167962-1G 167962-500MG 167962-5G
77465-688EA 668.47 USD
77465-688 77478-444 77488-632
1-Ethyl-4-(ethylthio)benzene ≥95%
1-Ethyl-4-(ethylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617717
CAS Number: 22905-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR