1-(Ethylthio)-3,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

167958-1G 167958-500MG 167958-5G
77465-684EA 648.9 USD
77465-684 77478-440 77488-628
1-(Ethylthio)-3,5-dimethylbenzene ≥95%
1-(Ethylthio)-3,5-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617713
CAS Number: 1108726-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR