1-(Ethylthio)-4-(mEthylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

167951-1G 167951-500MG 167951-5G 167951-25G
77465-680EA 258.53 USD
77465-680 77478-436 77488-624 77499-184
1-(Ethylthio)-4-(mEthylthio)benzene ≥95%
1-(Ethylthio)-4-(mEthylthio)benzene

Precaución: For research use only

Formula: C₉H₁₂S₂
MW: 184.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617698
CAS Number: 83500-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR