1-(1,1-Dimethylethyl)-4-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

167947-1G 167947-500MG 167947-5G
77465-676EA 668.47 USD
77465-676 77478-432 77488-620
1-(1,1-Dimethylethyl)-4-(ethylthio)benzene ≥95%
1-(1,1-Dimethylethyl)-4-(ethylthio)benzene

Precaución: For research use only

Formula: C₁₂H₁₈S
MW: 194.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617703
CAS Number: 30506-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR