1-(Ethylthio)-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

167943-1G 167943-500MG 167943-5G 167943-25G
77465-672EA 130.81 USD
77465-672 77478-428 77488-616 77499-180
1-(Ethylthio)-3-methoxybenzene ≥95%
1-(Ethylthio)-3-methoxybenzene

Precaución: For research use only

Formula: C₉H₁₂OS
MW: 168.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617684
CAS Number: 38474-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR