1-(Ethylthio)-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

167935-1G 167935-500MG 167935-5G 167935-25G
77465-668EA 106.09 USD
77465-668 77478-424 77488-612 77499-178
1-(Ethylthio)-3-methylbenzene ≥95%
1-(Ethylthio)-3-methylbenzene

Precaución: For research use only

Formula: C₉H₁₂S
Storage Temperature: Ambient
MDL Number: MFCD11617742
CAS Number: 34786-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR