4-Bromo-2-methoxy-5-methylbenzoic acid ≥96%

Supplier: AOBChem USA

Warning

167589-1G 167589-500MG 167589-5G
77465-660EA 479.98 USD
77465-660 77478-416 77488-604
4-Bromo-2-methoxy-5-methylbenzoic acid ≥96%
4-Bromo-2-methoxy-5-methylbenzoic acid

Precaución: For research use only

Formula: C₉H₉BrO₃
MW: 245.0699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852310
CAS Number: 2293347-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR