4-Chloro-2,6-diisopropylphenol ≥97%
Supplier: AOBChem USA![](https://pr.vwr.com/supplier/smallweb/Logo AOBChemUSA.jpg)
|
Warning
|
167499-1G
167499-500MG
167499-5G
77465-656EA
568.56
USD
77465-656
77478-412
77488-600
4-Chloro-2,6-diisopropylphenol ≥97%
4-Chloro-2,6-diisopropylphenol
Precaución: For research use only
Formula:
C₁₂H₁₇ClO MW: 212.7157 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24713587 CAS Number: 91561-75-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...