Ethyl 2-bromo-6-fluoro-α-hydroxybenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

167440-1G 167440-500MG 167440-5G
77465-652EA 546.93 USD
77465-652 77478-408 77488-596
Ethyl 2-bromo-6-fluoro-α-hydroxybenzeneacetate ≥95%
Ethyl 2-bromo-6-fluoro-α-hydroxybenzeneacetate

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₃
MW: 277.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852267
CAS Number: 1541312-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR