Ethyl 3-bromo-α-hydroxybenzeneacetate ≥96%

Supplier: AOBChem USA

Warning

167342-1G 167342-500MG 167342-5G 167342-25G
77465-648EA 338.87 USD
77465-648 77478-404 77488-592 77499-168
Ethyl 3-bromo-α-hydroxybenzeneacetate ≥96%
Ethyl 3-bromo-α-hydroxybenzeneacetate

Precaución: For research use only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD16693958
CAS Number: 62123-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR