Ethyl 2,4-dichloro-α-hydroxybenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

167304-1G 167304-500MG 167304-5G
77465-644EA 447.02 USD
77465-644 77478-400 77488-588
Ethyl 2,4-dichloro-α-hydroxybenzeneacetate ≥95%
Ethyl 2,4-dichloro-α-hydroxybenzeneacetate

Precaución: For research use only

Formula: C₁₀H₁₀Cl₂O₃
MW: 249.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16693963
CAS Number: 41204-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR