2-(6-Bromo-2-chloro-3-methylphenyl)-1,3-Dioxolane ≥95%

Supplier: AOBChem USA

Warning

16715-1G 16715-500MG 16715-5G 16715-25G
77465-640EA 327.54 USD
77465-640 77478-396 77488-584 77499-160
2-(6-Bromo-2-chloro-3-methylphenyl)-1,3-Dioxolane ≥95%
2-(6-Bromo-2-chloro-3-methylphenyl)-1,3-Dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrClO₂
MW: 277.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619379
CAS Number: 2221812-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR