3-Chloro-2-fluoro-6-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

167081-1G 167081-500MG 167081-5G
77465-636EA 1115.49 USD
77465-636 77478-392 77488-578
3-Chloro-2-fluoro-6-methylbenzoic acid ≥97%
3-Chloro-2-fluoro-6-methylbenzoic acid

Precaución: For research use only

Formula: C₈H₆ClFO₂
MW: 188.5834 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712075
CAS Number: 1427382-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR