α-Ethyl-2-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167071-1G 167071-500MG 167071-5G 167071-25G
77465-632EA 397.58 USD
77465-632 77478-388 77488-574 77499-154
α-Ethyl-2-(1-methylethyl)benzenemethanol ≥96%
α-Ethyl-2-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521230
CAS Number: 958810-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR