α-Ethyl-5-fluoro-2-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167062-1G 167062-500MG 167062-5G
77465-628EA 471.74 USD
77465-628 77478-384 77488-570
α-Ethyl-5-fluoro-2-methylbenzenemethanol ≥96%
α-Ethyl-5-fluoro-2-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₃FO
MW: 168.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521256
CAS Number: 1249487-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR