α-Ethyl-2-fluoro-5-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167059-1G 167059-500MG 167059-5G
77465-624EA 471.74 USD
77465-624 77478-380 77488-566
α-Ethyl-2-fluoro-5-methylbenzenemethanol ≥96%
α-Ethyl-2-fluoro-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD15478774
CAS Number: 1270584-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR