α-Ethyl-3-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167054-1G 167054-500MG 167054-5G
77465-620EA 648.9 USD
77465-620 77478-376 77488-562
α-Ethyl-3-(1-methylethyl)benzenemethanol ≥96%
α-Ethyl-3-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521231
CAS Number: 1314904-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR