α-Ethyl-4-methoxy-3,5-dimethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167051-1G 167051-500MG 167051-5G
77465-616EA 648.9 USD
77465-616 77478-372 77488-558
α-Ethyl-4-methoxy-3,5-dimethylbenzenemethanol ≥96%
α-Ethyl-4-methoxy-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD11521215
CAS Number: 1314932-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR