α-Ethyl-3-fluoro-5-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167049-1G 167049-500MG 167049-5G 167049-25G
77465-612EA 397.58 USD
77465-612 77478-368 77488-554 77499-150
α-Ethyl-3-fluoro-5-methylbenzenemethanol ≥96%
α-Ethyl-3-fluoro-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₃FO
MW: 168.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521247
CAS Number: 1314961-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR