α-Ethyl-4-(ethylthio)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

167035-1G 167035-500MG 167035-5G 167035-25G
77465-608EA 297.67 USD
77465-608 77478-364 77488-550 77499-148
α-Ethyl-4-(ethylthio)benzenemethanol ≥96%
α-Ethyl-4-(ethylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22004091
CAS Number: 1485337-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR