4-Bromo-3-chloro-α-ethyl-2-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167016-1G 167016-500MG 167016-5G
77465-604EA 447.02 USD
77465-604 77478-360 77488-546
4-Bromo-3-chloro-α-ethyl-2-fluorobenzenemethanol ≥95%
4-Bromo-3-chloro-α-ethyl-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34852123
CAS Number: 1691744-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR