3-Ethoxy-α-ethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

166989-1G 166989-500MG 166989-5G 166989-25G
77465-600EA 203.94 USD
77465-600 77478-356 77488-542 77499-144
3-Ethoxy-α-ethylbenzenemethanol ≥96%
3-Ethoxy-α-ethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O₂
MW: 180.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521249
CAS Number: 909567-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR