α-Ethyl-2,4-dimethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

166978-1G 166978-500MG 166978-5G 166978-25G
77465-596EA 314.15 USD
77465-596 77478-352 77488-538 77499-142
α-Ethyl-2,4-dimethylbenzenemethanol ≥96%
α-Ethyl-2,4-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11521214
CAS Number: 1216114-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR