4-Chloro-α-ethyl-2-fluorobenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

166962-1G 166962-500MG 166962-5G
77465-592EA 507.79 USD
77465-592 77478-348 77488-534
4-Chloro-α-ethyl-2-fluorobenzenemethanol ≥96%
4-Chloro-α-ethyl-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD17165114
CAS Number: 1427380-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR