3,6-Dibromo-α-ethyl-2-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166956-1G 166956-500MG 166956-5G 166956-25G
77465-588EA 415.09 USD
77465-588 77478-344 77488-530 77499-138
3,6-Dibromo-α-ethyl-2-fluorobenzenemethanol ≥95%
3,6-Dibromo-α-ethyl-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₉Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD34852112
CAS Number: 1628003-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR