α,4-Diethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166939-1G 166939-500MG 166939-5G 166939-25G
77465-584EA 350.2 USD
77465-584 77478-340 77488-526 77499-134
α,4-Diethylbenzenemethanol ≥95%
α,4-Diethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11521236
CAS Number: 74532-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR