α-Ethyl-3,5-dimethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166936-1G 166936-500MG 166936-5G 166936-25G
77465-580EA 295.61 USD
77465-580 77478-336 77488-522 77499-130
α-Ethyl-3,5-dimethoxybenzenemethanol ≥95%
α-Ethyl-3,5-dimethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O₃
MW: 196.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11835554
CAS Number: 203912-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR