2-Chloro-α-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166922-1G 166922-500MG 166922-5G 166922-25G
77465-576EA 394.49 USD
77465-576 77478-332 77488-516 77499-124
2-Chloro-α-ethylbenzenemethanol ≥95%
2-Chloro-α-ethylbenzenemethanol

Precaución: For research use only

Formula: C₉H₁₁ClO
MW: 170.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427115
CAS Number: 22869-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR