α-Ethyl-2-methoxybenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

166915-1G 166915-500MG 166915-5G 166915-25G
77465-572EA 394.49 USD
77465-572 77478-328 77488-512 77499-120
α-Ethyl-2-methoxybenzenemethanol ≥97%
α-Ethyl-2-methoxybenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427118
CAS Number: 7452-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR