α-Ethyl-α-methyl-3-(methylthio)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

166855-1G 166855-500MG 166855-5G 166855-25G
77465-568EA 339.9 USD
77465-568 77478-324 77488-508 77499-116
α-Ethyl-α-methyl-3-(methylthio)benzenemethanol ≥97%
α-Ethyl-α-methyl-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528421
CAS Number: 1379355-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR